Learn More
Medchemexpress LLC HY-B1768 100mg Medchemexpress, Succimer CAS:304-55-2 Purity:>98%
Supplier: Medchemexpress LLC HYB1768100MG
Medchemexpress, HY-B1768 100mg Succimer CAS:304-55-2 Succimer is a widely used chelating agent for the treatment of Pb poisoning. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.