Learn More
Chemglass Life Sciences Sublimer Cold Finger, Minum-Ware
Supplier: Chemglass Life Sciences MW7501
Sublimer Cold Finger, Minum-Ware. The long cold finger is inserted thru a MW-20 claisen adapter and into a MW-54 thin wall reaction vial and then filled with ice water. A MW-32 thermometer adapter is then inserted into the other arm of the claisen adapter and connected to a vacuum source. The vial is heated with a Bunsen burner, while the ice is replaced every 1-2 minutes with a Pasteur pipette. With the Bunsen burner, it is possible to chase the sublimate off the side walls of the vial. Sublimation takes place rapidly-within 5 minutes. Cold finger has a 14/10 inner joint. (Designed and developed by, Dr. Don Pavia and Ruth Schoonover, Western Washington University, Bellingham, Washington.)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.