Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #49.
Supplier: Strem, An Ascensus Company 96400050PC
50pcs. Suba-Seal #49. . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 24.0mm Dimensions A 25.2mm B 20.7mm Strem# 96-4000. http://www.strem.com/catalog/v/96-4000/