Learn More
Strem, An Ascensus Company 50pcs. Suba-Seal #37 (fits female 19/22 joint).
Supplier: Strem, An Ascensus Company 96350050PC
50pcs. Suba-Seal #37 (fits female 19/22 joint). . Molecular Weight: . Molecular Formula: . Color/form: Fits neck i.d. 19.0mm Dimensions A 20.4mm B 15.7mm Strem# 96-3500. http://www.strem.com/catalog/v/96-3500/