Learn More
World Precision Instrument SU-MPC325 Motorized Micromanipulator Systems-SU-MPC325-4
Supplier: World Precision Instrument SUMPC3254
The SU-MP225 represents an economical alternative to the SU-MP285 and SU- MPC365. In 2002, production and design changes allowed us to produce this motorized manipulator as a more affordable alternative to the industry standard SU-MP285. While the SU-MP225 feature set is less comprehensive than the SU-MP285, it includes the most popular features with an efficient user interface. The mechanical design utilizes a miniature stepper motor and integral anti-backlash gear head. Pre-loaded ball bearing slides provide smooth movement throughout the 25 mm of travel. The controller uses low-noise, linear-drive output circuitry identical to that found in the SU-MP285. The methodology for mounting pipette holders and headstages used with the MP-285 has been maintained in the SU-MP225 to allow for cross compatibility.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.