Learn More
World Precision Instrument SU-MP85 Micromanipulator-SU-MP85-L
Supplier: World Precision Instrument SUMP85L
The classic micromanipulator developed by Sir Huxley many years ago is still considered by many investigators to be the finest manual micromanipulator available. The SU-MP85 offers the advantage of a very large range of movement with its built-in coarse manipulator and precise submicron movement with the fine controls. Coarse positioning is achieved with a three axis manipulator mounted on top. The ultrafine positioning is accomplished with the large micrometers mounted on the base. These micrometers provide smooth, precise movement through a 10:1 reduction mechanism.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.