Learn More
GSC Go Science Crazy Student Distillation Apparatus Kit
Demonstrates the properties of distillation in an inclusive kit. The GSC International Student Distillation Apparatus Kit is a must have kit for any student interested in learning the properties of distillation. It includes everything needed for a distillation demonstration.
Supplier: GSC Go Science Crazy 4002
Alert:
Intended for ages 13 and up.
Includes: Liebig condenser w rubber connections, 300mm length; Kjeldahl ball, 50mm diameter; Boiling flask, 500mL; Three legged support stand for boiling flask; Cast iron support stand; Wire gauze; 500mL Erlenmeyer flask; Instructions
Description
- Simple and inexpensive pieces contained in this distilling apparatus are perfect for a student set
- Contains everything necessary to demonstrate the properties of distillation
- A comprehensive booklet is included which explains how the process of distillation is used in industry
- Boiling Flask, Erlenmeyer Flask and Kjeldahl Ball are made from heat resistant borosilicate glass
- Sizes: Liebig Condenser - 300mm L; Kjeldahl Ball - 50mm D; Boiling Flask - 500mL; Erlenmeyer Flask - 500mL
Specifications
Demonstrating properties of distillation | |
Borosilicate glass, cast iron, rubber |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov