Learn More
Cole-Parmer Stuart RE3022C Diaphragm Vacuum Pump; 230 VAC
Supplier: Cole-Parmer 9996696
Use this compact diaphragm vacuum pump for chemical laboratory vacuum generation, especially well suited to rotary evaporation. Vacuum regulator with gauge and adjustment knob allows you to create ideal vacuum conditions in the lab. Oilless pump with integrated moisture protection is low maintenance.