Learn More
Research Products International Corp Mini-Strip Blotting Box, 1 Lane, 11.1 x 5.2 x 1.4cm, 5 per Package
SDP
Supplier:  Research Products International Corp 248775
Encompass_Preferred
Perfect size container for incubating or staining western blot strips. Clear plastic box has individual chambers that allow processing multiple strips at one time. Flat bottom on container reduces the amount of solution necessary to cover each strip. Box also features a tight fitting hinged lid that prevents evaporation. Made of high clarity polystyrene for enhanced visibility of contents.
Catalog No. 50-550-310