Learn More
Thermo Scientific™ Streptex™ Latex Agglutination Test
Isolate and identify a variety of streptococci with a simple enzyme extraction method that detects Lancefield groups A, B, C, D, F and G.
$1050.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Streptex Latex Agglutination Test |
CE Marker | Yes |
Type | Streptococcus Test Kit |
For Use With (Application) | Enzyme Immunoassay Detection |
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R30950501
|
Thermo Scientific™
R30950501 |
Streptex Latex Agglutination Test | 50 Tests/Kit | 2°C to 8°C |
Pack of 50 for $1,050.00
|
|
|||
Description
Rapidly identify and differentiate Lancefield group of streptococci using the Thermo Scientific™ Streptex™ Latex Agglutination Test. The test utilizes latex particles sensitized with group-specific antibodies which agglutinate in presence of homologous antigens.
- Rely on tried-and-true tests with more than 50 years of combined proven performance
- Maintain confidence with reliable results that have greater than 99% sensitivity (according to data available in the package inserts)
- Clear-cut, robust reactions provide easy-to-read results
- Improve productivity with compact kit configurations, color-coded reagents and convenient workstations
- Flexible use from selective and non-selective media
- Control latex is not required
- Maximize spending efficiency with the option of purchasing separate kit components
- Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Specifications
2°C to 8°C | |
Yes | |
Enzyme Immunoassay Detection | |
Polyvalent |
Streptex Latex Agglutination Test | |
Streptococcus Test Kit | |
Test latex, extraction enzyme, polyvalent positive control, disposable reaction cards, disposable mixing sticks | |
Lancefield A, B, C, D, F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.