Learn More
Streck Retic-Chex™ II Controls
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Retic-Chex II is a tri-level whole blood control with reticulocytes used to verify the accuracy and precision of manual reticulocyte counting methods.
Supplier: Streck 285212
This item is not returnable.
View return policy
Description
- Can be used for manual and automated reticulocyte counting.
- Available in three clinically significant levels.
- Packaged in 1.0 mL plastic squeeze dropper vials for accurate dispensing.
- 14-day open-vial stability, 75-day closed-vial stability.
- Established assay ranges provide less room for error.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Retic-Chex II Configuration Level 1 and 2 | |
Hematology II Controls |