Learn More
Streck Pipet Labs Calibration Kits
Verify automatic pipets and dilutors without purchasing separate equipment or interrupting pipet use
$93.00
Specifications
Product Type | Maintenance Kit |
---|---|
For Use With (Application) | To perform simple titrations, analysis and calculation of the results |
Quantity | Each |
Includes: Six vials of receiver solution and one or two vials of pipetting solution
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
23-042-701
![]() |
Streck
271279 |
201 to 5000 μL |
Each for $93.00
|
|
23-042-700
![]() |
Streck
271278 |
10 to 200 μL |
Each for $93.00
|
|
Description
- Kit allows user to perform six simple titrations
- Prepaid return postage
- Emailed report sent within 48 hours
Order Info
Order one PVS kit per pipet to be tested
Specifications
Maintenance Kit | |
To perform simple titrations, analysis and calculation of the results | |
Each |