Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Para 4 Hematology Controls
Available in plastic dropper vials for easy sample allocation with hemoglobin and hematocrit instruments
$115.00
Specifications
Certifications/Compliance | CE |
---|---|
Content And Storage | 2°C to 10°C |
Format | Plastic Vial |
For Use With (Equipment) | HemoCue™ B-Hemoglobin photometer and Hb 201+ analyzer, HemataSTAT II™ |
Type | Para 4™ Hematology Control |
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-285-631
|
Streck
215106 |
6 Low |
Each for $115.00
|
|
|||||
23-285-633
|
Streck
215108 |
6 High |
Each for $115.00
|
|
|||||
23-285-632
|
Streck
215107 |
6 Normal |
Each for $115.00
|
|
|||||
Description
- Assay includes red blood cell (RBC), white blood cell (WBC), Hgb, Hct and platelet values for auxiliary methods
- 110-day closed-vial stability
- 14-day open-vial stability
- HemoCue™ B-Hemoglobin Photometer and Hb 201+ analyzer
- Separation Technology, Inc. HemataSTAT II™
Specifications
CE | |
Plastic Vial | |
Para 4™ Hematology Control | |
6 x 1.5 mL | |
110 Days |
2°C to 10°C | |
HemoCue™ B-Hemoglobin photometer and Hb 201+ analyzer, HemataSTAT II™ | |
For evaluating the accuracy and precision of automated, semi automated and manual procedures that measure Blood cell parameters | |
Stabilized Human RBC, a White Blood Cell Component Consisting of Human and/or Non-human aAnalogs and a Platelet Component |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title