Learn More
Streck ESR-Vacuum Tubes
ESR-Vacuum Tubes for use with the Streck ESR-Auto Plus™ and ESR-100 Sedimentation Rate Analyzers.
Supplier: Streck 240320
This item is not returnable.
View return policy
Description
Collect, transport, and analyze the Erythrocyte Sedimentation Rate (ESR) of whole blood in the clinical laboratory with Streck ESR-Vacuum Tubes. They are specifically designed for use with the Streck ESR-Auto Plus and ESR-100 automated sedimentation rate analyzers and the Streck ESR-10 Manual Rack.
- For the Collection and Analysis of Whole Blood
- Extended Stability: Patient samples can be maintained from the time of blood collection for up to 72 hours prior to analysis when transported and stored at 2-10°C in all Streck ESR-Vacuum Tubes
- Tubes contain tri-sodium citrate to avoid coagulation
- After the tube is mixed, it is placed in the ESR instrument; the instrument automatically prints or downloads the ESR result in 30 minutes
- Significantly more impact resistant than glass collection tubes
- Designed with an outer Mylar™ coating that contains glass and blood specimens in the event of breakage
- Reduces the risk of injury and potential exposure to blood borne pathogens
Specifications
ESR Vacuum Tube | |
Streck ESR-Auto Plus™ and ESR-100 Sedimentation Rate Analyzers | |
Tri Sodium citrate to avoid coagulation | |
30 min. |
2 mL | |
100 Box | |
2°C to 10°C |