Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streamlight™ ProPolymer™ 4AA LED
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Ergonomically shaped for comfort
Supplier: Streamlight™ 68201
Includes: Wrist lanyard connection
Description
- Seven super-bright, 100,000-hour LEDs provide up to 155 hours of continuous use (27 lumens typical with seven white LEDs)
- Shock-resistant, engineering-grade yellow polymer case with unbreakable polycarbonate lens
- Waterproof, nonconductive, and corrosion-proof
Order Info
Powered by four AA alkaline batteries. Length: 6.5 in. (16.5cm). Weight: 6.3 oz. (179g).
Certifications
UL listed for certain Class I, Division 1 hazardous locations; CE and MSHA marked.
Specifications
Flashlight | |
6.3 oz. | |
155 hr. | |
CE and MSHA, UL Class I, Div. 1, Groups C, D; Class I, Div 2, Groups A, B, C, D; Class II, Div. 2, Groups F&G; Class III, T3C | |
Wrist lanyard connection |
White LED | |
0.173 kg | |
Alkaline | |
Yellow or Black | |
67 lm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction