Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Streak Collection
Demonstrates identifying properties.
Supplier: American Educational Products 2361
Includes: Streak plate
Description
Some minerals leave a colored streak on unglazed tile, providing a clue to identifying that mineral. Examples of this property can be demonstrated with this collection.
- Eight number-coded specimens
- Specimen size: 1 × 1.5 in. (2.5 × 3.8cm)
Specifications
Streak plate to identify minerals | |
8 Specimens | |
2.5 x 3.8 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction