Learn More
Perkin Elmer US LLC Storm Gas Cell - Stainless Steel, 10 cm, KBr
Supplier: Perkin Elmer US LLC L1272475
Short Path Length Storm Gas Cells are ideal for routine gas and vapor spectroscopic analysis. They are available in either pyrex or stainless steel, with either NaCl or KBr windows. These cells require, but do not include the Universal 10 cm Gas Cell Mount (51000817).