Learn More
Cole-Parmer Oakton Conductivity Probe Storage Solution; 500 mL
Supplier: Cole-Parmer 0065280
Conductivity Probe Storage Solution is ideal for storing and conditioning your conductivity cells. It preserves the probe's cleanliness, allows immediate use, and reduces contamination that can occur with storing probe in water only.