Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Supplier: Axygen™ PDW12_HC
Description
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.Specifications
ANSI, SLAS 1-2004 | |
Polypropylene | |
Non-sterile | |
Round | |
50/Cs. | |
-112°F |
Clear | |
96 | |
1.2 mL | |
Sample Collection, in-Vitro Growth Chambers and Long Term Storage | |
5/Pk. | |
-80°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction