Learn More
Ace Glass, Inc. Stopper, impinger, tube end inlet/outlets, 24/25, 0.035incapillary tip
SDP
Supplier:  Ace Glass, Inc. 754106
Encompass_Preferred
24/25 impinger stopper with tube end inlet/outlets and a 0.035in capillary tip that is positioned 4mm above the bottom of the bottle. Stopper fits 24/25, 125mL bottle and features a 11mm O.D. center tube, and a 8mm O.D. side arm with bulge.
Catalog No. 50-979-326