Learn More
Ace Glass, Inc. Stopper, impinger, 12/5 socket inlet, 12/5 ball outlet, 24/40, calibrated nozzle
Supplier: Ace Glass, Inc. 753128
24/40 midget impinger stopper with 12/5 socket inlet and 12/5 ball outlet. Features a calibrated nozzle that delivers 0.09-0.11 CFM at 30.5cm vacuum.