Learn More
Revvity Health Sciences Inc STK-Substrate 3-biotin, 50 g
Supplier: Revvity Health Sciences Inc 61ST3BLE
This substrate is a component of HTRF KinEASE S3 kit developed for an optimal kinase enzymatic step. It can be ordered as a spare part, e.g. when studying serine/threonine kinases with exceptionally high substrate Km.