Learn More
Fisherbrand™ Stirring Paddles
Ideal for use in stirring small volumes or for chemical mixing in photographic work.
$41.00 - $130.00
Specifications
For Use With (Application) | Stirring small volumes or for chemical mixing in photographic work |
---|
Catalog Number | Height (English) | Length (English) | Size | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Height (English) | Length (English) | Size | Width (English) | Price | Quantity & Availability | |||
S37117
|
0.52 in. | 21.9 in. | Small | 2 in. |
Case of 4 for $60.25
|
|
|||
S37118
|
40.5 in. | 3 in. | Large | 1.02 in. |
Case of 4 for $130.00
|
|
|||
S37119
|
76.2 in. | 3 in. | Large | 1.02 in. |
Each for $41.00
|
|
|||
Description
Choose from small or large size paddles.
- Made from high-density polyethylene
- Small paddle is ideal for use in stirring small volumes or for chemical mixing in photographic work
- Large paddles are excellent for mixing liquids in large tanks or vats
Order Info
Small paddle handle dia. is 0.5 in. (1.3 cm); large paddle handle dia. is 1 in. dia. (2.5 cm). Additional stirring paddle sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Stirring small volumes or for chemical mixing in photographic work |