Learn More
Ampco Safety Tools Stike Box Wrench, 12-pt (Imperial)
AMPCO™ Strike Wrench, 12pt Box performs in high-risk and demanding environments.
$317.50 - $10560.00
Specifications
Product Type | Wrench Strike 12pt Box |
---|---|
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
19-174-877
|
Ampco Safety Tools
WS1 |
1 in. |
Each for $327.50
|
|
|||||
19-174-880
|
Ampco Safety Tools
WS11/16 |
11/16 in. |
Each for $317.50
|
|
|||||
19-174-882
|
Ampco Safety Tools
WS11/2 |
1-1/2 in. |
Each for $482.50
|
|
|||||
19-174-883
|
Ampco Safety Tools
WS11/4 |
1-1/4 in. |
Each for $414.50
|
|
|||||
19-174-884
|
Ampco Safety Tools
WS11/8 |
1-1/8 in. |
Each for $365.50
|
|
|||||
19-174-886
|
Ampco Safety Tools
WS111/16 |
1-11/16 in. |
Each for $482.50
|
|
|||||
19-174-887
|
Ampco Safety Tools
WS113/16 |
1-13/16 in. |
Each for $508.00
|
|
|||||
19-174-891
|
Ampco Safety Tools
WS13/16 |
1-3/16 in. |
Each for $414.50
|
|
|||||
19-174-892
|
Ampco Safety Tools
WS13/4 |
1-3/4 in. |
Each for $482.50
|
|
|||||
19-174-893
|
Ampco Safety Tools
WS13/8 |
1-3/8 in. |
Each for $424.50
|
|
|||||
19-174-897
|
Ampco Safety Tools
WS15/16 |
1-5/16 in. |
Each for $414.50
|
|
|||||
19-174-898
|
Ampco Safety Tools
WS15/8 |
1-5/8 in. |
Each for $482.50
|
|
|||||
19-174-902
|
Ampco Safety Tools
WS17/16 |
1-7/16 in. |
Each for $441.00
|
|
|||||
19-174-903
|
Ampco Safety Tools
WS17/8 |
1-7/8 in. |
Each for $558.00
|
|
|||||
19-174-919
|
Ampco Safety Tools
WS19/16 |
1-9/16 in. |
Each for $482.50
|
|
|||||
19-174-920
|
Ampco Safety Tools
WS2 |
2 in. |
Each for $558.00
|
|
|||||
19-174-921
|
Ampco Safety Tools
WS21/16 |
2-1/16 in. |
Each for $583.50
|
|
|||||
19-174-922
|
Ampco Safety Tools
WS21/2 |
2-1/2 in. |
Each for $748.00
|
|
|||||
19-174-923
|
Ampco Safety Tools
WS21/4 |
2-1/4 in. |
Each for $731.50
|
|
|||||
19-174-924
|
Ampco Safety Tools
WS21/8 |
2-1/8 in. |
Each for $600.00
|
|
|||||
19-174-925
|
Ampco Safety Tools
WS213/16 |
2-13/16 in. |
Each for $1,053.00
|
|
|||||
19-174-926
|
Ampco Safety Tools
WS215/16 |
2-15/16 in. |
Each for $1,078.00
|
|
|||||
19-174-928
|
Ampco Safety Tools
WS23/16 |
2-3/16 in. |
Each for $600.00
|
|
|||||
19-174-929
|
Ampco Safety Tools
WS23/4 |
2-3/4 in. |
Each for $1,037.00
|
|
|||||
19-174-930
|
Ampco Safety Tools
WS23/8 |
2-3/8 in. |
Each for $748.00
|
|
|||||
19-174-932
|
Ampco Safety Tools
WS25/16 |
2-5/16 in. |
Each for $748.00
|
|
|||||
19-174-933
|
Ampco Safety Tools
WS25/8 |
2-5/8 in. |
Each for $1,018.00
|
|
|||||
19-174-936
|
Ampco Safety Tools
WS27/16 |
2-7/16 in. |
Each for $748.00
|
|
|||||
19-174-939
|
Ampco Safety Tools
WS29/16 |
2-9/16 in. |
Each for $748.00
|
|
|||||
19-174-940
|
Ampco Safety Tools
WS3 |
3 in. |
Each for $1,178.00
|
|
|||||
19-174-941
|
Ampco Safety Tools
WS3/4 |
3/4 in. |
Each for $317.50
|
|
|||||
19-174-944
|
Ampco Safety Tools
WS31/2 |
3-1/2 in. |
Each for $2,420.00
|
|
|||||
19-174-945
|
Ampco Safety Tools
WS31/4 |
3-1/4 in. |
Each for $1,178.00
|
|
|||||
19-174-946
|
Ampco Safety Tools
WS31/8 |
3-1/8 in. |
Each for $1,178.00
|
|
|||||
19-174-949
|
Ampco Safety Tools
WS33/4 |
3-3/4 in. |
Each for $2,470.00
|
|
|||||
19-174-950
|
Ampco Safety Tools
WS33/8 |
3-3/8 in. |
Each for $2,330.00
|
|
|||||
19-174-953
|
Ampco Safety Tools
WS35/8 |
3-5/8 in. |
Each for $2,470.00
|
|
|||||
19-174-956
|
Ampco Safety Tools
WS37/8 |
3-7/8 in. |
Each for $2,470.00
|
|
|||||
19-174-958
|
Ampco Safety Tools
WS4 |
4 in. |
Each for $3,190.00
|
|
|||||
19-174-961
|
Ampco Safety Tools
WS41/8 |
4-1/8 in. |
Each for $3,190.00
|
|
|||||
19-174-962
|
Ampco Safety Tools
WS411/16 |
4-11/16 in. |
Each for $6,660.00
|
|
|||||
19-174-963
|
Ampco Safety Tools
WS415/16 |
4-15/16 in. |
Each for $6,660.00
|
|
|||||
19-174-967
|
Ampco Safety Tools
WS45/16 |
4-5/16 in. |
Each for $3,190.00
|
|
|||||
19-174-971
|
Ampco Safety Tools
WS49/16 |
4-9/16 in. |
Each for $6,660.00
|
|
|||||
19-174-974
|
Ampco Safety Tools
WS51/8 |
5-1/8 in. |
Each for $6,660.00
|
|
|||||
19-174-975
|
Ampco Safety Tools
WS511/16 |
5-11/16 in. |
Each for $10,560.00
|
|
|||||
19-174-977
|
Ampco Safety Tools
WS53/8 |
5-3/8 in. |
Each for $10,560.00
|
|
|||||
19-174-983
|
Ampco Safety Tools
WS6 |
6 in. |
Each for $10,560.00
|
|
|||||
19-174-989
|
Ampco Safety Tools
WS7/8 |
7/8 in. |
Each for $320.50
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Wrench Strike 12pt Box | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.