Learn More
CPC Scientific H-Leu-Thr-Glu-Arg-His-Lys-Ile-Leu-His-Arg-Leu-Leu-Gln-Glu-OH (trifluoroacetate salt) 5MG
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CPC Scientific SIGN014B
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This peptide is a 14-amino acid fragment from the steroid receptor cofactor SRC-1 NR IIONE-LETTER SEQUENCE: LTERHKILHRLLQEMOLECULAR FORMULA: C79H136N26O21MOLECULAR WEIGHT:1786.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: InflammationREFERENCES: Bramlett, K. et al. Mol. Endocrinol. 15, 909 (2001)