Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Sterile Syringe Filter PES
Used for clarifying solutions.
$262.00
Specifications
Material (Membrane) | PES |
---|---|
Sterility | Sterile |
Housing Material | Acrylic |
Inlet Style | Female Luer-Lok |
Outlet Style | Male Leur-slip |
Catalog Number | Pore Size | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Pore Size | Quantity | Price | Quantity & Availability | |||||
09-720-514
|
0.45 μm | 50/Pk. |
Each for $262.00
|
|
|||||
09-720-511
|
0.22 μm | 50/Pk. |
Each for $262.00
|
|
|||||
Description
Sterile filtration of protein solutions, tissue culture media, additives, buffers, and waterIndividually wrapped
Specifications
PES | |
Acrylic | |
Male Leur-slip | |
33 mm |
Sterile | |
Female Luer-Lok | |
50/Pk. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title