Learn More
STEMCELL Technologies Culture Tubes, 14 mL, Size: 500 Units
Supplier: STEMCELL Technologies 2000522
Use sterile, polystyrene Culture Tubes, 14mL for a wide variety of sample preparation, storage, and centrifugation tasks. These high-quality round-bottom tubes are recommended for EasySep™ cell separation and have been verified for use with "The Big Easy" EasySep™ Magnet (Catalog #18001) or EasyEights™ EasySep™ Magnet (14 mL side; Catalog #18103). These tubes can also be used for flow cytometry and fluorescence-activated cell sorting (FACS). These culture tubes are equivalent to Corning Catalog #352057. For more information, visit www.stemcell.com/cultureware.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.