Learn More
STEMCELL Technologies Culture Tubes, 5 mL, Size: 500 Units
Supplier: STEMCELL Technologies 2000523
Use sterile, polypropylene Culture Tubes, 5mL for a wide variety of sample preparation, storage, and centrifugation tasks. These high-quality round-bottom tubes are recommended for select protocols in cell isolation, flow cytometry, and fluorescence-activated cell sorting (FACS). These culture tubes are equivalent to Corning Catalog #352063. For more information, visit www.stemcell.com/cultureware.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.