Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Sterile Disposable Petri Dishes with Covers
Eisco™ Sterile Disposable Petri Dishes with Covers made from transparent and nontoxic polystyrene are ready to use, individually packaged in polythylene bags and sterilized by radiation.
Supplier: Eisco™ CH0374C
Description
- Made from polystyrene
- Transparent
- Includes dish and cover
- Individually packaged
- Sterilized via radiation
- Ready to use
- Disposable
Warranty
1 year limited manufacturer warranty
Specifications
Transparent | |
Polystyrene | |
Sterile | |
Culturing | |
15 mm |
Single-use | |
Round | |
Radiation | |
1 | |
35 mm |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'