Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Sterile Cell Strainer PROMO
Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50 mL conical tubes. Available in three color-coded mesh sizes.
This item is not returnable.
View return policy
Description
Rapidly isolate primary cells
- Fast, simple alternative to gauze filtration
- Reliably dissociates cells from clumps or primary tissues
- Consistently obtain a uniform single-cell suspension
- Ideal for preparation of:
- Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
- Specimens for primary cell cultures and immunogens
- Freezing stocks
- Filtering agglutinative proteins produced in inactivation serum
Reliable and easy to use
- Strong nylon mesh with evenly spaced pores for consistent results
- Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
- Includes an extended lip for easier handling with forceps
- Color-coded polypropylene frame with tab enables easy handling and identification
- Individually packed, ready to use and disposable
Specifications
Nylon | |
White | |
Cell Strainer | |
Conical Tubes, 50 mL |
Single-use | |
Polypropylene | |
Sterile | |
70 μm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction