Learn More
World Precision Instrument Steri-Lite Bead Sterilizer-ST5191-AU
Supplier: World Precision Instrument ST5191AU
WPI's Steri-Lite Glass Bead Sterilizer is ideal for the sterilization of small research instruments (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms.