Learn More
Instech STER LOCK SOLN HEP 5/10ML W/PP
Supplier: Instech USPHDS50010VYP5
Pharmaceutical-grade catheter lock solution prepared with heparin sodium following the USP <797> standard for compounding sterile preparations. The solution contains heparin (500 USP units/ml) in 50% dextrose, five 10ml vials, yellow, PinPort™ access.