Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Public Seating™ Steel Height Adjustable Science Lab Table with HPL Top
Built to last in an industrial environment
Supplier: National Public Seating™ SLT32472H
Description
- 16 gauge steel frame apron and 14 gauge legs
- 11 gauge steel tabletop support tabs and corner brackets fuse as one with four heavy-duty tamper-free Loctite hex screws
- Adjustable height steel legs feature a supreme dual connection
- Leg-base is fastened to tabletop with four heavy-duty screws which gets inserted into the frame using a strong welded hangar bolt
- Legs connect to frame corner brackets with industrial-grade nylon-insert lock nuts
- Legs adjust in 1 in. increments and lock firmly in place with two K-9 external tooth lock washers and heavy duty tamper-free hex screws
- All legs come standard with simple-adjust glides with optional casters
- High Pressure Laminate table top provides the ideal surface for lab experiments
Specifications
Science Lab Table | |
Black | |
72 x 24 x 22.25 to 37.25 in. | |
HPL |
MAS2 | |
Industrial Use | |
22.25 to 37.25 in. | |
10 years |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction