Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate 98.0+%, TCI America™
SDP

Catalog No. D1644100G
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 2082-79-3
Molecular Formula C35H62O3
Molecular Weight (g/mol) 530.878
MDL Number MFCD00017498
InChI Key SSDSCDGVMJFTEQ-UHFFFAOYSA-N
Synonym 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester, Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
PubChem CID 16386
IUPAC Name octadecyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 53°C
Color White
Quantity 100 g
Formula Weight 530.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
EINECSNumber (3)-1737
TSCA Yes
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.