Learn More
Yamato Scientific America, Inc. Yamato 20L Steam Sterilizer with Dryer, 220V
Supplier: Yamato Scientific America, Inc. SM211
High performance, fully automatic 20L standard steam sterilization from start to finish with high pressure steam sterilization and drying steps. Automatic operations from sterilization to drying carried out with an interactive key input system. Quick sample drying capability makes samples ready to use right after sterilization. Drying temperature can be set according to sample material, quantity, etc. Timer range from 1~999 hours. Drain bottle water level can be quickly confirmed on the front panel level indicator. Drain valve located in front for easy access. Absence of protrusions in sterilization chamber makes insertion & removal of baskets, and other items quick and easy. Self-diagnostic functions make operation safer and error recovery quicker. Condensation collector neutralizes high temperature exhaust steam safely. Sterilizing temp. range 105 to 123°C. Drying temp range 150 to 180°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.