Learn More
Cytiva LevMixer System
Supplier: Cytiva 74030998S
LevMixer System is the ideal component for product homogenization, suspension or re-suspension, final formulation and media, buffer preparation. LevMixer takes advantage of technology capable of suspension powers and other solids without clogging or shearing. The system is mobile and flexible to fit all users' spatial needs while offering a wide range of mixing volumes for a broad series of applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.