Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Status H. Pylori Test Kit
SDP

Rapid immunoassay for the qualitative detection of the anti-helicobacter pylori antibody in human whole blood, serum or plasma

Supplier:  LifeSign 37030

Encompass

Catalog No. 23-264-796


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Features:

  • CLIA waived for whole blood
  • Sensitivity of 95.9% with 91.6% overall agreement to biopsy histology
  • Easy-to-use single reagent procedure
  • Results available in 10 minutes
  • Requires only 25μL of whole blood or 10μL serum/plasma
  • Convenient fingerstick sample collection
Specifications

Specifications

91.6% overall agreement to biopsy histology
Whole Blood, Serum, Plasma
30 Tests
Anti-helicobacter pylori antibodies
H. pylori Immunoassay
10 min.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit