Learn More
Static Solutions™ OHM-Shield™ AF-6000 Zero VOC ESD Floor Finish
SDP
Combines static protection, durability and gloss with environmentally friendly chemicals.
Supplier:  Static Solutions™ AF6005
Encompass
Catalog No. 19-130-6033
Add to Cart
Description
Description
Protects the floor with a long lasting and durable gloss without harming the environment. No buffing required. Produces zero VOCs (Volatile Organic Compounds); eliminates the possibility of outgassing. Safe for your workers as well as your piece parts. Can be used in closed environments.
  • High gloss
  • Zinc-free
  • Biodegradable
  • Low odor
  • Static dissipative
  • No dye or fragrance
  • High percentage of solids (18-20%)
Order Info
Resistance: 107 to 109 ohms/sq. Static Dissipation: 0.02 seconds. Color: Translucent white; dries clear. Coverage: 1500-2500 sq. ft. per gallon. Note: Allow 1-2 hours of drying time between coats (2-3 recommended). Coated floor should be earth grounded for rapid dissipation. Store tightly closed in a cool dry area, and do not mix other floor finishes or left over products from previous applications.
Certifications
UL listed for slip resistance.
Compliance
EOS/ESD S202-20 compliant. RoHS compliant. Exceeds EPA guidelines.
Specifications
Specifications
Zero VOC ESD Floor Finish
Medical, electronics and cleanroom environments, this product protects the floor with a long lasting and durable gloss
18.9 L
OHM-Shield™
5 gal.
Product Suggestions
Product Suggestions