Learn More
StatLab™ StatCut™ Microtome Blades
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Suitable for both soft and hard tissue, StatCut™ blades are well balanced in durability and sharpness.
Supplier: StatLab™ CUT7950
This item is not returnable.
View return policy
Description
- Blades are coated for non-stick sections and effortless ribboning
Specifications
High | |
Single-use | |
50 Pk. |