Learn More
EKF Diagnostics STAT-Site™ Whole Blood β-Ketone Control Solution, Box foil pack
Whole blood β-ketone measurement in seconds.
Supplier:  EKF Diagnostics 7020100
Catalog No. 22-022-653
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • 1 μL sample size
  • Uses capillary or venous whole blood
  • Box foil pack, 100 pcs.
  • Hand-held, easy to use
Specifications
Specifications
Test Strips