Learn More
Axygen™ Starter Kit 1
Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
Supplier: Axygen™ APSTRKIT1
Description
The Axygen Axypet Starter Kit contains: AP-20, AP-200, and AP-1000 pipettors; 1 rack each of 200 μL and 1000 μL Axygen tips; calibration tool; ejector spacers set; identification labels; lubricant; color identification rings; right-hand ejector button.
- Cat. No. AP-20 Axygen Axypet single channel Pipettor 2 to 20 μL
- Cat. No. AP-200 Axygen Axypet single channel Pipettor 20 to 200 μL
- Cat. No. AP-1000 Axygen Axypet single channel Pipettor 100 to 1,000 μL
- 1 rack each of 200 μL and 1000 μL Axygen tips
Specifications
Axypet | |
Starter Kit | |
AP-20, AP-200, and AP-1000 pipettors; 1 rack each of 200 μL and 1000 μL Axygen tips; calibration tool; ejector spacers set; identification labels; lubricant; color identification rings; right-hand ejector button | |
1 Kit |