Learn More
Electron Microscopy Sciences Aurion ImmunoGold Starter UltraSmall Kit
Supplier: Electron Microscopy Sciences 2555010
Immunogold reagent kit. Ultra-Small Starter Kit consists of 100 ulUltra-small Conjugate, 10ml BSA-c, 2x2.5ml R-Gent SE-LM and a sample pack R-Gent SE-EM