Learn More
Starplex™ Scientific Specimen Containers: Cups
Supplier: Starplex™ Scientific B1202SYN
Description
- Break- and leak-resistant for safety during harsh transport conditions or pneumatic tube systems
- Clarified polypropylene cups provide easy visual analysis of sample
- Full-circle thread design prevents cross-threading
- Graduated markings allow quick measurement of sample
- Labeling provides ample space for patient information
- Ridges on cap provide a secure grip for safe handling
Specifications
Polypropylene | |
120 mL | |
Spout Cap |
Non-sterile | |
Yellow |