Learn More
Heidolph™ StarFish™ Glassware - Condensers
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Heidolph™ StarFish™ Glassware - Condensers are available in various lengths and joint sizes
$200.00 - $1429.00
Catalog Number | Mfr. No. | Type | Length (Metric) | Cone Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Length (Metric) | Cone Size | Price | Quantity & Availability | |||
05-404-107
![]() |
Heidolph™
015890800 |
Allihn | 250 mm | 19/26 |
Each for $1,429.00
|
|
|||
05-404-108
![]() |
Heidolph™
015890810 |
Allihn | 250 mm | 24/29 |
Each for $1,392.00
|
|
|||
05-404-105
![]() |
Heidolph™
015890780 |
Coil | 165 mm | 19/26 |
Each for $322.00
|
|
|||
05-404-106
![]() |
Heidolph™
015890790 |
Coil | 165 mm | 24/29 |
Each for $317.00
|
|
|||
05-404-103
![]() |
Heidolph™
015890760 |
Liebig | 208 mm | 19/26 |
Each for $200.00
|
|
|||
05-404-104
![]() |
Heidolph™
015890770 |
Liebig | 250 mm | 24/29 |
Each for $241.00
|
|
|||
Description
- Different types of connections available
- Condensers can be used for all standard distillations
- Available in various lengths/cone sizes
Specifications
1/Pk. |