Learn More
3M™ Petrifilm™ Staph Express Count Plate, Sold by AquaPhoenix Scientific
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Provides Staphylococcus aureus results
Supplier: 3M™ Petrifilm™ 6491
Description
- Results in 24 hours
- Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
- Provides accurate, efficient, consistent results for food processors and contract labs
- Unique system to facilitates enumeration of S. aureus
- S. aureus appears as distinctive red-violet colonies
- This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: 3M-01/9-04/03 A and 3M-01/9-04/03 B for All human foods and Pet food by AFNOR Certification
Note:
- Other colonies have to be checked using Staph express disk
Specifications
Staphylococcus aureus | |
Count Plate | |
Food processors and contract labs, Facilitating enumeration of S. aureus | |
500 Pack |