Learn More
QA Bio Inc QA-Bio, Inc. • MAb4 Antibody Reference Panel • 10 µgs
Supplier:  QA Bio Inc SAMAB4
Encompass
MAb4 Antibody Reference Panel • The Mab4 antibody reference panel is a mixture of glycans commonly found on monoclonal antibodies • includes G2F(NA2F), G0F(NGA2F), and both G1F isomers • 10 µg
Catalog No. 50-355-863