Learn More
Electron Microscopy Sciences Vitrobot™ Filter Paper, Grade 595, 50 mm
Supplier: Electron Microscopy Sciences 7116665
This special Filter Paper has an outer diameter of 55 mm, with an inner diameter of 20 mm. Made from Grade 595 paper.