Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 615
Fast flowing filter paper used as an intermediate grade between Grade 617 and 615. It has a rough surface and is used for filtering aluminum, ferric and magnesium hydroxides. It is also found in general construction filtration work and in the filtration of insoluble iron and ore.
Supplier: Ahlstrom-Munksjo 61500750
Specifications
25 μm | |
Filtering aluminum, ferric, magnesium hydroxides. For construction filtration work, filtration of insoluble iron and ore. | |
7.5 cm |
Fast | |
615 |