Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ QuStick™ Strep A Kit
For diagnostic use
$251.00 - $404.00
Specifications
Content And Storage | Room Temperature |
---|---|
Clia Complexity | Waived |
Detectable Analytes | Streptococcal Antigen (Group A) |
Sample Type | Throat Swab |
Sensitivity | 97% |
Includes: Test strips, reagents, controls, throat swabs, extraction tubes, instructions
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-022-515
|
Stanbio™
6000025 |
25 Tests |
Each for $251.00
|
|
|||||
22-022-516
|
Stanbio™
6000050 |
50 Tests |
Each for $404.00
|
|
|||||
Description
- Sensitivity: 97% compared to culture
- Results in 5 minutes
Certifications
CLIA waived
Specifications
Room Temperature | |
Streptococcal Antigen (Group A) | |
97% | |
Test strip, reagents, control, throat swabs, extraction tubes and instructions |
Waived | |
Throat Swab | |
Streptococcus A Test Kit | |
0.5 min. |
Safety and Handling
ShelfLife : 2 Years
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title