Learn More
Stanbio™ Quicktest RPR
Carbon flocculation slide test
$116.00 - $342.00
Specifications
Detectable Analytes | Reagin (Syphilis) |
---|---|
Includes | RPR Antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control |
Includes: RPR antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
SB-1170-100
|
Stanbio™
1170100 |
100 Tests | QuickTest RPR Test |
Pack of 100 for $116.00
|
|
||||
SB-1175-305
|
Stanbio™
1175305 |
3 x 5 mL | QuickTest RPR Test Control |
Each for $158.00
|
|
||||
SB-1170-500
|
Stanbio™
1170500 |
500 Tests | QuickTest RPR Test |
Pack of 500 for $342.00
|
|
||||
Description
- For qualitative and quantitative determination of reagin antibodies in serum or plasma
- Macroscopic non-treponemal flocculation test that is used to detect and quantify reagin, an anti-lipoidal antibody found in serum or plasma from persons with syphilis and occasionally in persons with acute or chronic infections or conditions other than syphilis
- Antigen is modification of VDRL antigen which contains microparticulate charcoal to enhance the visual difference between a reactive and non-reactive result
Specifications
Reagin (Syphilis) | |
RPR Antigen suspension, cardiolipin suspension, RPR reactive control, RPR weakly-reactive control, RPR non-reactive control |